Anti-Bisphospho-ERK1 pThr202/Tyr205 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA513036
CAPIPA5-13036EA 3164.49 CAD
CAPIPA5-13036
Anti-Bisphospho-ERK1 pThr202/Tyr205 Rabbit Polyclonal Antibody
Antibodies
This antibody is predicted to react with canine, mouse and rat based on sequence homology. MAPK3 is a member of the MAP kinase family. MAP kinases, also known as extracellular signal-regulated kinases (ERKs), act in a signaling cascade that regulates various cellular processes such as proliferation, differentiation, and cell cycle progression in response to a variety of extracellular signals. This kinase is activated by upstream kinases, resulting in its translocation to the nucleus where it phosphorylates nuclear targets.

Recommended Dilutions: Western Blot: 1:1000, Dot blot: 1:500

Type: Primary
Antigen: Bisphospho-ERK1 pThr202/Tyr205
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR