Anti-MAP2K1 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA513041
CAPIPA5-13041EA 2573.22 CAD
CAPIPA5-13041
Anti-MAP2K1 Rabbit Polyclonal Antibody
Antibodies
This antibody is predicted to react with chicken, canine, hamster, mouse, rat, rabbit and Xenopus based on sequence homology. MEK1is a dual specificity protein kinase that belongs to the MAP kinase kinase family. This kinase is known to play a critical role in mitogen growth factor signal transduction. It phosphorylates and thus activates MAPK1/ERK2 and MAPK2/ERK3. The activation of this kinase itself is dependent on the Ser/Thr phosphorylation by MAP kinase kinase kinases. The inhibition or degradation of this kinase is found to be involved in the pathogenesis of Yersinia and anthrax.

Recommended Dilutions: Western Blot: 1:500, Dot blot: 1:500

Type: Primary
Antigen: MAP2K1 (mitogen-activated protein kinase kinase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR