Anti-HIPK2 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA513045
CAPIPA5-13045EA 2801.79 CAD
CAPIPA5-13045
Anti-HIPK2 Rabbit Polyclonal Antibody
Antibodies
This antibody is predicted to react with mouse and rat based on sequence homology. HIPK2 is a conserved serine/threonine nuclear kinase that interacts with homeodomain transcription factors.

Recommended Dilutions: Dot blot: 1:500

Type: Primary
Antigen: HIPK2 (homeodomain interacting protein kinase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR