Anti-FABP4 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA513052
CAPIPA5-13052EA 2573.22 CAD
CAPIPA5-13052
Anti-FABP4 Rabbit Polyclonal Antibody
Antibodies
This antibody is predicted to react with bovine, mouse, porcine and rat based on sequence homology. FABP4 is a fatty acid binding protein found in adipocytes. Fatty acid binding proteins are a family of small, highly conserved, cytoplasmic proteins that bind long-chain fatty acids and other hydrophobic ligands. It is thought that FABPs roles include fatty acid uptake, transport, and metabolism.

Recommended Dilutions: Dot blot: 1:500

Type: Primary
Antigen: FABP4 (fatty acid binding protein 4, adipocyte)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR