Anti-CLDN2 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA513068
CAPIPA5-13068EA 2573.22 CAD
CAPIPA5-13068
Anti-CLDN2 Rabbit Polyclonal Antibody
Antibodies
Members of the claudin protein family, such as CLDN2, are expressed in an organ-specific manner and regulate the tissue-specific physiologic properties of tight junctions.

Recommended Dilutions: Dot blot: 1:500

Type: Primary
Antigen: CLDN2 (claudin 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR