Anti-SOX2 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA513080
CAPIPA5-13080EA 2286.72 CAD
CAPIPA5-13080
Anti-SOX2 Rabbit Polyclonal Antibody
Antibodies
This antibody is predicted to react with mouse based on sequence homology. SOX2 is a member of the SRY-related HMG-box (SOX) family of transcription factors involved in the regulation of embryonic development and in the determination of cell fate. This protein may act as a transcriptional activator after forming a protein complex with other proteins. Mutations in the SOX2 gene have been associated with bilateral anophthalmia, a severe form of structural eye malformation.

Recommended Dilutions: Dot blot: 1:500

Type: Primary
Antigen: SOX2 (SRY (sex determining region Y)-box 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR