Anti-ABCB5 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA513114
CAPIPA5-13114EA 2801.79 CAD
CAPIPA5-13114
Anti-ABCB5 Rabbit Polyclonal Antibody
Antibodies
ABCB5 belongs to the ATP-binding cassette (ABC) transporter superfamily of integral membrane proteins. These proteins participate in ATP-dependent transmembrane transport of structurally diverse molecules ranging from small ions, sugars, and peptides to more complex organic molecules.

Flow Cytometry: 1:10-1:50, Western Blot: 1:1000

Type: Primary
Antigen: ABCB5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR