Anti-CDX1 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA513121
CAPIPA5-13121EA 2561.45 CAD
CAPIPA5-13121
Anti-CDX1 Rabbit Polyclonal Antibody
This gene is a member of the caudal-related homeobox transcription factor gene family. The encoded DNA-binding protein regulates intestine-specific gene expression and enterocyte differentiation. It has been shown to induce expression of the intestinal alkaline phosphatase gene, and inhibit beta-catenin/T-cell factor transcriptional activity.

Immunohistochemistry: 1:50-1:100, Western Blot: 1:1000

Type: Primary
Antigen: CDX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR