Anti-S1PR1 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA513127
CAPIPA5-13127EA 2573.22 CAD
CAPIPA5-13127
Anti-S1PR1 Rabbit Polyclonal Antibody
Antibodies
EDG1 is structurally similar to G protein-coupled receptors and is highly expressed in endothelial cells. It binds the ligand sphingosine-1-phosphate with high affinity and high specificity, and suggested to be involved in the processes that regulate the differentiation of endothelial cells. Activation of this receptor induces cell-cell adhesion.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: S1PR1 (sphingosine-1-phosphate receptor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR