Anti-S1PR3 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA513129
CAPIPA5-13129EA 2801.79 CAD
CAPIPA5-13129
Anti-S1PR3 Rabbit Polyclonal Antibody
Antibodies
EDG3 is a member of the EDG family of receptors, which are G protein-coupled receptors. This protein has been identified as a functional receptor for sphingosine 1-phosphate and likely contributes to the regulation of angiogenesis and vascular endothelial cell function.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: S1PR3 (sphingosine-1-phosphate receptor 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR