Anti-LPAR2 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA513130
CAPIPA5-13130EA 2444.36 CAD
CAPIPA5-13130
Anti-LPAR2 Rabbit Polyclonal Antibody
This antibody is predicted to react with non-human primate based on sequence homology. EDG4 is a member of family I of the G protein-coupled receptors, as well as the EDG family of proteins. This protein functions as a lysophosphatidic acid (LPA) receptor and contributes to Ca2+ mobilization, a critical cellular response to LPA in cells, through association with Gi and Gq proteins.

Recommended Dilutions: Western Blot: 1:1000, Immunohistochemistry: 1:50-1:100

Type: Primary
Antigen: LPAR2 (lysophosphatidic acid receptor 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR