Anti-ICOS Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA513135
CAPIPA5-13135EA 2801.79 CAD
CAPIPA5-13135
Anti-ICOS Rabbit Polyclonal Antibody
Antibodies
ICOS belongs to the CD28 and CTLA-4 cell-surface receptor family. It forms homodimers and plays an important role in cell-cell signaling, immune responses, and regulation of cell proliferation.

Recommended Dilutions: Western Blot: 1:1000, Immunohistochemistry: 1:50-1:100, Flow Cytometry: 1:10-1:50

Type: Primary
Antigen: ICOS (inducible T-cell co-stimulator)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR