Anti-MAGEA6 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA513154
CAPIPA5-13154EA 2561.45 CAD
CAPIPA5-13154
Anti-MAGEA6 Rabbit Polyclonal Antibody
MAGEA6 is a member of the MAGEA gene family. The members of this family have their entire coding sequences located in the last exon, and the encoded proteins show 50 to 80% sequence identity between each other. The promoters and first exons of the MAGEA genes show considerable variability, suggesting that the existence of this gene family enables the same function to be expressed under different transcriptional controls. The MAGEA genes are expressed at a high level in a number of tumors of various histologic types, and are silent in normal tissues with the exception of testis and placenta. The MAGEA genes are clustered on chromosome Xq28. They may be implicated in some hereditary disorders, such as dyskeratosis congenita.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: MAGEA6 (melanoma antigen family A, 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR