Anti-MMP11 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA513180
CAPIPA5-13180EA 2444.36 CAD
CAPIPA5-13180
Anti-MMP11 Rabbit Polyclonal Antibody
Proteins of the matrix metalloproteinase (MMP) family are involved in the breakdown of extracellular matrix in normal physiological processes, such as embryonic development, reproduction, and tissue remodeling, as well as in disease processes, such as arthritis and metastasis. Most MMPs are secreted as inactive proproteins which are activated when cleaved by extracellular proteinases. MMP11 may play an important role in the progression of epithelial malignancies. It is specifically expressed in stromal cells of breast carcinomas.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: MMP11 (matrix metallopeptidase 11 (stromelysin 3))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR