Anti-MMP20 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA513188
CAPIPA5-13188EA 2286.08 CAD
CAPIPA5-13188
Anti-MMP20 Rabbit Polyclonal Antibody
Antibodies
MMP20 degrades amelogenin, the major protein component of the enamel matrix, as well as two of the macromolecules characterizing the cartilage extracellular matrix: aggrecan and the cartilage oligomeric matrix protein (COMP). MMP20 may play a central role in tooth enamel formation.

Recommended Dilutions: Western Blot: 1:1000, Immunohistochemistry: 1:50-1:100

Type: Primary
Antigen: MMP20 (matrix metallopeptidase 20)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR