Anti-OASL Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA513213
CAPIPA5-13213EA 2561.45 CAD
CAPIPA5-13213
Anti-OASL Rabbit Polyclonal Antibody
2-prime,5-prime oligoadenylates (2-5As) bind to and activate RNase L, leading to degradation of RNA and inhibition of protein synthesis. 2-5As are produced by 2-5A synthetases (OASs), a highly-conserved family of interferon-induced enzymes. The predicted 514-amino acid human p59OASL (2-5A synthetases-like) protein shares a highly conserved N-terminal domain with other OASs. The C-terminal portion of p59OASL contains 2 ubiquitin-like domains. p59OASL is expressed in most tissues, with the highest levels in hematopoietic tissues, colon, and stomach.

Recommended Dilutions: Western Blot: 1:1000, Immunohistochemistry: 1:50-1:100

Type: Primary
Antigen: OASL (2'-5'-oligoadenylate synthetase-like)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR