Anti-SMYD5 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA513217
CAPIPA5-13217EA 2573.22 CAD
CAPIPA5-13217
Anti-SMYD5 Rabbit Polyclonal Antibody
Antibodies
Retinoic acid plays a critical role in development, cellular growth, and differentiation. The specific function of the protein for this intronless, retinoic acid-induced gene has not yet been determined; however, it has been suggested to play a role in development.

Recommended Dilutions: Western Blot: 1:1000, Immunohistochemistry: 1:50-1:100

Type: Primary
Antigen: SMYD5 (SMYD family member 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR