Anti-GPRC5A Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA513221
CAPIPA5-13221EA 2561.45 CAD
CAPIPA5-13221
Anti-GPRC5A Rabbit Polyclonal Antibody
Retinoic acid plays a critical role in development, cellular growth, and differentiation. The specific function for the protein from this retinoic acid-induced gene has not yet been determined; however, it may play a role in embryonic development and epithelial cell differentiation. Rai3 is a member of the type 3 G protein-coupling receptor family, characterized by the signature 7-transmembrane domain motif. It may also be involved in interaction between retinoid acid and G protein signalling pathways.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: GPRC5A (G protein-coupled receptor, class C, group 5, member A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR