Anti-SIRT7 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA513226
CAPIPA5-13226EA 2286.72 CAD
CAPIPA5-13226
Anti-SIRT7 Rabbit Polyclonal Antibody
Antibodies
This antibody is predicted to react with bovine, mouse and rat based on sequence homology. SIRT7 is a member of the sirtuin family of proteins, homologs to the yeast Sir2 protein. Members of the sirtuin family are characterized by a sirtuin core domain and grouped into four classes. The functions of human sirtuins have not yet been determined; however, yeast sirtuin proteins are known to regulate epigenetic gene silencing and suppress recombination of rDNA. Studies suggest that the human sirtuins may function as intracellular regulatory proteins with mono-ADP-ribosyltransferase activity.

  • Recommended Dilutions: Western Blot: 1:1000, Immunohistochemistry: 1:50-1:100

Type: Primary
Antigen: SIRT7 (sirtuin 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR