Anti-Cdc25A Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA513259
CAPIPA5-13259EA 2561.45 CAD
CAPIPA5-13259
Anti-Cdc25A Rabbit Polyclonal Antibody
CDC25A is a member of the CDC25 family of phosphatases. CDC25A is required for progression from G1 to the S phase of the cell cycle. It activates the cyclin-dependent kinase CDC2 by removing two phosphate groups. CDC25A is specifically degraded in response to DNA damage, which prevents cells with chromosomal abnormalities from progressing through cell division. CDC25A is an oncogene, although its exact role in oncogenesis has not been demonstrated.

Western Blot: 1:1000

Type: Primary
Antigen: Cdc25A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR