Anti-EDA Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA513276
CAPIPA5-13276EA 2231.48 CAD
CAPIPA5-13276
Anti-EDA Rabbit Polyclonal Antibody
This antibody is predicted to react with bovine based on sequence homology. EDA is a type II membrane protein that can be cleaved by furin to produce a secreted form. This protein, which belongs to the tumor necrosis factor family, acts as a homotrimer and may be involved in cell-cell signaling during the development of ectodermal organs. Defects in the gene for EDA are a cause of ectodermal dysplasia, anhidrotic, which is also known as X-linked hypohidrotic ectodermal dysplasia.

Recommended Dilutions: Western Blot: 1:1000, Immunohistochemistry: 1:10-1:50, Flow Cytometry: 1:10-1:50

Type: Primary
Antigen: EDA (ectodysplasin A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR