Anti-GRB2 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA513281
CAPIPA5-13281EA 2573.22 CAD
CAPIPA5-13281
Anti-GRB2 Rabbit Polyclonal Antibody
Antibodies
This antibody is predicted to react with mouse, rat and Xenopus based on sequence homology. GRB2 binds the epidermal growth factor receptor and contains one SH2 domain and two SH3 domains. Its two SH3 domains direct complex formation with proline-rich regions of other proteins, and its SH2 domain binds tyrosine phosphorylated sequences.

Recommended Dilutions: Western Blot: 1:1000, Immunohistochemistry: 1:50-1:100

Type: Primary
Antigen: GRB2 (growth factor receptor-bound protein 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR