Anti-INA Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA513283
CAPIPA5-13283EA 2561.45 CAD
CAPIPA5-13283
Anti-INA Rabbit Polyclonal Antibody
This antibody is predicted to react with bovine, mouse and rat based on sequence homology. INA is a class-IV neuronal intermediate filament that is able to self-assemble. It is involved in the morphogenesis of neurons. It may form an independent structural network without the involvement of other neurofilaments or it may cooperate with NF-L to form the filamentous backbone to which NF-M and NF-H attach to form the cross-bridges.

Immunohistochemistry: 1:10-1:50, Western Blot: 1:1000

Type: Primary
Antigen: INA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR