Anti-CD14 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA513305
CAPIPA5-13305EA 2984.36 CAD
CAPIPA5-13305
Anti-CD14 Rabbit Polyclonal Antibody
Antibodies
CD14 is a surface protein preferentially expressed on monocytes/macrophages. It binds lipopolysaccharide binding protein and recently has been shown to bind apoptotic cells.

Flow Cytometry: 1:10-1:50, Immunocytochemistry: 1:10-1:50, Immunofluorescence: 1:10-1:50, Immunohistochemistry (Paraffin): 1:10-1:50, Western Blot: 1:1000

Type: Primary
Antigen: CD14
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR