Anti-PARP3 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA513308
CAPIPA5-13308EA 2801.79 CAD
CAPIPA5-13308
Anti-PARP3 Rabbit Polyclonal Antibody
Antibodies
PARP3, poly(ADP-ribosyl)transferase 3, belongs to the PARP family. These enzymes modify nuclear proteins by poly-ADP-ribosylation, which is required for DNA repair, regulation of apoptosis, and maintenance of genomic stability. This protein is preferentially localized to the daughter centriole throughout the cell cycle.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: PARP3 (poly (ADP-ribose) polymerase family, member 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR