Anti-CLDN1 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA513332
CAPIPA5-13332EA 2561.45 CAD
CAPIPA5-13332
Anti-CLDN1 Rabbit Polyclonal Antibody
Tight junctions represent one mode of cell-to-cell adhesion in epithelial or endothelial cell sheets, forming continuous seals around cells and serving as a physical barrier to prevent solutes and water from passing freely through the paracellular space. These junctions are comprised of sets of continuous networking strands in the outwardly facing cytoplasmic leaflet, with complementary grooves in the inwardly facing extracytoplasmic leaflet. CLDN1, a member of the claudin family, is an integral membrane protein and a component of tight junction strands. Loss of function mutations result in neonatal ichthyosis-sclerosing cholangitis syndrome.

Immunohistochemistry: 1:10-1:50, Western Blot: 1:1000

Type: Primary
Antigen: CLDN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR