Anti-RCAN3 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA513336
CAPIPA5-13336EA 2814.74 CAD
CAPIPA5-13336
Anti-RCAN3 Rabbit Polyclonal Antibody
DSCR1L2 inhibits calcineurin-dependent transcriptional responses by binding to the catalytic domain of calcineurin A, and may play a role during central nervous system development. Highest expression occurs in heart, skeletal muscle kidney, liver and peripheral blood leukocytes.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: RCAN3 (RCAN family member 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR