Anti-SPPL3 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA513338
CAPIPA5-13338EA 1896.83 CAD
CAPIPA5-13338
Anti-SPPL3 Rabbit Polyclonal Antibody
Signal peptide peptidase (SPP) is an aspartyl protease that mediates clearance of signal peptides by proteolysis within the endoplasmic reticulum (ER). Like presenilins, SPP contains a critical GXGD motif in its C-terminal catalytic center. SPPL3 is one of several presenilin homologues/SPP-like proteins (PSHs/SPPL) that have been identified.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: SPPL3 (signal peptide peptidase like 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR