Anti-KLK5 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA513345
CAPIPA5-13345EA 2250.89 CAD
CAPIPA5-13345
Anti-KLK5 Rabbit Polyclonal Antibody
Kallikreins are a subgroup of serine proteases having diverse physiological functions. Growing evidence suggests that many kallikreins are implicated in carcinogenesis and some have potential as novel cancer and other disease biomarkers. KLK5 expression is up-regulated by estrogens and progestins. The protein is secreted and may be involved in desquamation in the epidermis.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: KLK5 (kallikrein-related peptidase 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR