Anti-CLCN4 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA513348
CAPIPA5-13348EA 2561.45 CAD
CAPIPA5-13348
Anti-CLCN4 Rabbit Polyclonal Antibody
This antibody is predicted to react with mouse and rat based on sequence homology. The CLCN family of voltage-dependent chloride channel genes comprises nine members (CLCN1-7, Ka and Kb) which demonstrate quite diverse functional characteristics while sharing significant sequence homology. Chloride channel 4 has an evolutionary conserved CpG island and is conserved in both mouse and hamster. This gene is mapped in close proximity to APXL (Apical protein Xenopus laevis-like) and OA1 (Ocular albinism type I), which are both located on the human X chromosome at band p22.3. The physiological role of chloride channel 4 remains unknown but may contribute to the pathogenesis of neuronal disorders.

Immunocytochemistry: 1:10-1:50, Immunofluorescence: 1:10-1:50, Immunohistochemistry (Frozen): 1:10-1:50, Western Blot: 1:1000

Type: Primary
Antigen: CLCN4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR