Anti-CSF2RB Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA513380
CAPIPA5-13380EA 2561.45 CAD
CAPIPA5-13380
Anti-CSF2RB Rabbit Polyclonal Antibody
Purified rabbit polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide. This antibody is first purified by protein G affinity chromatography. Then, the antibody fraction is peptide affinity purified in a 2-step procedure with the control and phosphorylated peptides. The phospho-specific antibody is eluted with high and low pH buffers and neutralized immediately, followed by dialysis against PBS.

Western Blot: 1:1000

Type: Primary
Antigen: CSF2RB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR