Anti-PARP1 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA513386
CAPIPA5-13386EA 2286.72 CAD
CAPIPA5-13386
Anti-PARP1 Rabbit Polyclonal Antibody
Antibodies
This antibody is predicted to react with mouse based on sequence homology. PARP1 is a chromatin-associated enzyme, poly(ADP-ribosyl)transferase, which modifies various nuclear proteins by poly(ADP-ribosyl)ation. The modification is dependent on DNA and is involved in the regulation of various important cellular processes such as differentiation, proliferation, and tumor transformation and also in the regulation of the molecular events involved in the recovery of cell from DNA damage. In addition, this enzyme may be the site of mutation in Fanconi anemia, and may participate in the pathophysiology of type I diabetes.

Recommended Dilutions: Western Blot: 1:1000, Immunofluorescence: 1:10-1:50, Flow Cytometry: 1:10-1:50

Type: Primary
Antigen: PARP1 (poly (ADP-ribose) polymerase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR