Anti-HDGF Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA513389
CAPIPA5-13389EA 2561.45 CAD
CAPIPA5-13389
Anti-HDGF Rabbit Polyclonal Antibody
HDGF is a member of the hepatoma-derived growth factor family. This protein has mitogenic and DNA-binding activity and may play a role in cellular proliferation and differentiation.

Recommended Dilutions: Western Blot: 1:1000, Immunofluorescence: 1:10-1:50, Immunocytochemistry: 1:10-1:50, Immunohistochemistry: 1:10-1:50

Type: Primary
Antigen: HDGF (hepatoma-derived growth factor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR