Anti-FGF1 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA513394
CAPIPA5-13394EA 2801.79 CAD
CAPIPA5-13394
Anti-FGF1 Rabbit Polyclonal Antibody
Antibodies
This antibody is predicted to react with porcine based on sequence homology. FGF1 is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. This protein functions as a modifier of endothelial cell migration and proliferation, as well as an angiogenic factor. It acts as a mitogen for a variety of mesoderm- and neuroectoderm-derived cells in vitro, thus is thought to be involved in organogenesis.

Recommended Dilutions: Western Blot: 1:1000, Immunofluorescence: 1:10-1:50, Immunohistochemistry: 1:10-1:50

Type: Primary
Antigen: FGF1 (fibroblast growth factor 1 (acidic))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR