Anti-FXN Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA513410
CAPIPA5-13410EA 2801.79 CAD
CAPIPA5-13410
Anti-FXN Rabbit Polyclonal Antibody
Antibodies
This antibody is predicted to react with mouse based on sequence homology. FXN is a mitochondrial protein which belongs to the FRATAXIN family. The protein functions in regulating mitochondrial iron transport and respiration. The expansion of intronic trinucleotide repeat GAA results in Friedreich ataxia.

Recommended Dilutions: Western Blot: 1:1000, Immunohistochemistry: 1:10-1:50

Type: Primary
Antigen: FXN (frataxin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR