Anti-FXN Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA513411
CAPIPA5-13411EA 2286.08 CAD
CAPIPA5-13411
Anti-FXN Rabbit Polyclonal Antibody
Antibodies
FXN is a mitochondrial protein which belongs to the FRATAXIN family. The protein functions in regulating mitochondrial iron transport and respiration. The expansion of intronic trinucleotide repeat GAA results in Friedreich ataxia.

Recommended Dilutions: Western Blot: 1:1000, Immunofluorescence: 1:10-1:50, Immunocytochemistry: 1:10-1:50, Immunohistochemistry: 1:10-1:50

Type: Primary
Antigen: FXN (frataxin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR