Anti-EDNRA Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA513432
CAPIPA5-13432EA 2573.22 CAD
CAPIPA5-13432
Anti-EDNRA Rabbit Polyclonal Antibody
Antibodies
This antibody is predicted to react with mouse and rat based on sequence homology. EDNRA is a receptor for endothelin-1. Mediates its action by association with G proteins that activate a phosphatidylinositol-calcium second messenger system.

Recommended Dilutions: Western Blot: 1:1000, Immunofluorescence: 1:10-1:50, Flow Cytometry: 1:10-1:100

Type: Primary
Antigen: EDNRA (endothelin receptor type A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR