Anti-SUV420H2 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA513436
CAPIPA5-13436EA 2801.79 CAD
CAPIPA5-13436
Anti-SUV420H2 Rabbit Polyclonal Antibody
Antibodies
This antibody is predicted to react with rat based on sequence homology. SUV420H2 is a histone methyltransferase that specifically trimethylates 'Lys-20' of histone H4. H4 'Lys-20' trimethylation represents a specific tag for epigenetic transcriptional repression. The protein mainly functions in pericentric heterochromatin regions, thereby playing a central role in the establishment of constitutive heterochromatin in these regions. SUV420H1 is targeted to histone H3 via its interaction with RB1 family proteins (RB1, RBL1 and RBL2).

Recommended Dilutions: Western Blot: 1:1000, Flow Cytometry: 1:10-1:50

Type: Primary
Antigen: SUV420H2 (suppressor of variegation 4-20 homolog 2 (Drosophila))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR