Anti-CEA Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA513438
CAPIPA5-13438EA 2801.79 CAD
CAPIPA5-13438
Anti-CEA Rabbit Polyclonal Antibody
Antibodies
CEA is a cell surface glycoprotein that plays a role in cell adhesion and in intracellular signaling. It is a receptor for E.coli Dr adhesins.

Flow Cytometry: 1:10-1:50, Immunohistochemistry (Paraffin): 1:50-1:100, Western Blot: 1:1000

Type: Primary
Antigen: CEA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR