Anti-FLI1 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA513440
CAPIPA5-13440EA 2801.79 CAD
CAPIPA5-13440
Anti-FLI1 Rabbit Polyclonal Antibody
Antibodies
This antibody is predicted to react with bovine, chicken, canine, rat and Xenopus and zebrafish based on sequence homology. FLI1 is a sequence-specific transcriptional activator. It recognizes the DNA sequence 5'-C[CA]GGAAGT-3'.

Recommended Dilutions: Western Blot: 1:1000, Immunofluorescence: 1:10-1:50, Immunocytochemistry: 1:10-1:50

Type: Primary
Antigen: FLI1 (Fli-1 proto-oncogene, ETS transcription factor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR