Anti-ARPC1A Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA513448
CAPIPA5-13448EA 2561.45 CAD
CAPIPA5-13448
Anti-ARPC1A Rabbit Polyclonal Antibody
This antibody is predicted to react with bovine, mouse and rat based on sequence homology. ARPC1A is one of seven subunits of the human Arp2/3 protein complex. This subunit is a member of the SOP2 family of proteins and is most similar to the protein ARPC1B. The similarity between these two proteins suggests that they both may function as p41 subunit of the human Arp2/3 complex that has been implicated in the control of actin polymerization in cells. It is possible that the p41 subunit is involved in assembling and maintaining the structure of the Arp2/3 complex. Multiple versions of the p41 subunit may adapt the functions of the complex to different cell types or developmental stages.

Flow Cytometry: 1:10-1:50, Immunohistochemistry: 1:50-1:100, Western Blot: 1:1000

Type: Primary
Antigen: ARPC1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR