Anti-CLNS1A Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA513449
CAPIPA5-13449EA 2444.36 CAD
CAPIPA5-13449
Anti-CLNS1A Rabbit Polyclonal Antibody
This antibody is predicted to react with rabbit based on sequence homology. CLNS1A is a protein that functions in multiple regulatory pathways. The protein complexes with numerous cytosolic proteins and performs diverse functions including regulation of small nuclear ribonucleoprotein biosynthesis, platelet activation and cytoskeletal organization. The protein is also found associated with the plasma membrane where it functions as a chloride current regulator.

Flow Cytometry: 1:10-1:50, Immunohistochemistry: 1:50-1:100, Western Blot: 1:1000

Type: Primary
Antigen: CLNS1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR