Anti-CNOT8 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA513451
CAPIPA5-13451EA 2801.79 CAD
CAPIPA5-13451
Anti-CNOT8 Rabbit Polyclonal Antibody
Antibodies
This antibody is predicted to react with mouse based on sequence homology. Ubiquitous transcription factor required for a diverse set of processes. The CCR4-NOT complex functions as general transcription regulation complex.

Flow Cytometry: 1:10-1:50, Immunocytochemistry: 1:10-1:50, Immunofluorescence: 1:10-1:50, Immunohistochemistry: 1:10-1:50, Western Blot: 1:1000

Type: Primary
Antigen: CNOT8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR