Anti-ENO1 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA513459
CAPIPA5-13459EA 2801.79 CAD
CAPIPA5-13459
Anti-ENO1 Rabbit Polyclonal Antibody
Antibodies
This antibody is predicted to react with bovine, chicken, non-human primate, rat and Xenopus based on sequence homology. ENO1 is one of three enolase isoenzymes found in mammals; it is alpha-enolase, a homodimeric soluble enzyme, and also a shorter monomeric structural lens protein, tau-crystallin. The two proteins are made from the same message. The full length protein, the isoenzyme, is found in the cytoplasm. The shorter protein is produced from an alternative translation start, is localized to the nucleus, and has been found to bind to an element in the c-myc promoter.

Recommended Dilutions: Western Blot: 1:1000, Immunofluorescence: 1:10-1:50, Immunocytochemistry: 1:10-1:50, Immunohistochemistry: 1:10-1:50, Flow Cytometry: 1:10-1:50

Type: Primary
Antigen: ENO1 (enolase 1, (alpha))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR