Anti-EPN3 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA513460
CAPIPA5-13460EA 2561.45 CAD
CAPIPA5-13460
Anti-EPN3 Rabbit Polyclonal Antibody
This antibody is predicted to react with mouse and rat based on sequence homology. EPN3 belongs to the epsin family.

Recommended Dilutions: Western Blot: 1:1000, Immunohistochemistry: 1:10-1:50, Flow Cytometry: 1:10-1:50

Type: Primary
Antigen: EPN3 (epsin 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR