Anti-HFE Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA513482
CAPIPA5-13482EA 2444.36 CAD
CAPIPA5-13482
Anti-HFE Rabbit Polyclonal Antibody
This antibody is predicted to react with rat based on sequence homology. HFE is a membrane protein that is similar to MHC class I-type proteins and associates with beta2-microglobulin (beta2M). It is thought that this protein functions to regulate iron absorption by regulating the interaction of the transferrin receptor with transferrin. The iron storage disorder, hereditary haemochromatosis, is a recessive genetic disorder that results from defects in its gene.

Recommended Dilutions: Western Blot: 1:1000, Immunohistochemistry: 1:50-1:100

Type: Primary
Antigen: HFE (hemochromatosis)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR