Anti-AMER1 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA513492
CAPIPA5-13492EA 2801.79 CAD
CAPIPA5-13492
Anti-AMER1 Rabbit Polyclonal Antibody
Antibodies
WTX is involved in kidney development.

Flow Cytometry: 1:10-1:50, Immunohistochemistry (Paraffin): 1:50-1:100, Western Blot: 1:1000

Type: Primary
Antigen: AMER1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR