Anti-CPXM2 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA513500
CAPIPA5-13500EA 2444.36 CAD
CAPIPA5-13500
Anti-CPXM2 Rabbit Polyclonal Antibody
CPXM2 may be involved in cell-cell interactions.

Western Blot: 1:1000

Type: Primary
Antigen: CPXM2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR