Anti-SERPINB2 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA513501
CAPIPA5-13501EA 2561.45 CAD
CAPIPA5-13501
Anti-SERPINB2 Rabbit Polyclonal Antibody
PAI2 inhibits urokinase-type plasminogen activator. The monocyte derived PAI-2 is distinct from the endothelial cell-derived PAI-1.

Recommended Dilutions: Western Blot: 1:1000, Immunohistochemistry: 1:10-1:50

Type: Primary
Antigen: SERPINB2 (serpin peptidase inhibitor, clade B (ovalbumin), member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR