Anti-DONSON Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA513509
CAPIPA5-13509EA 2561.45 CAD
CAPIPA5-13509
Anti-DONSON Rabbit Polyclonal Antibody
DONSON is essential for DNA amplification in the ovary and required for cell proliferation during development.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: DONSON (downstream neighbor of SON)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR